Gene Transcripts

Gene Symbol
Siah3
Entrez Gene ID
692004
Gene Name
siah E3 ubiquitin protein ligase family member 3
Gene Aliases
Chromosome Location
Chr. 15: 57505449 - 57568743 on Build Rnor_6.0
UniGene ID
Rn.228206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191118.1 NP_001178047.1 2 510

View Full Product Details