Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMED9
Entrez Gene ID
701114
Gene Name
transmembrane p24 trafficking protein 9
Gene Aliases
-
Chromosome Location
Chr.6: 175092836 - 175096987 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193984.2 NP_001180913.1 - 612 77
GenBank mRNA JU328176.1 - - 499 77
JV047446.1 - - 594 77
JV565140.1 - - 568 77

View Full Product Details