Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM7A
Entrez Gene ID
711830
Gene Name
lysine demethylase 7A
Gene Aliases
JHDM1D
Chromosome Location
Chr.3: 166506495 - 166601008 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015135046.2 XP_014990532.1 - 1433 112
GenBank mRNA JU326136.1 - - 1322 112
JV724219.1 - - 1234 112

View Full Product Details