Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLEC10
Entrez Gene ID
701960
Gene Name
collectin subfamily member 10
Gene Aliases
EGK_19229
Chromosome Location
Chr.8: 119353115 - 119393241 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145915.2 XP_015001401.1 - 247 94

View Full Product Details