Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYAL4
Entrez Gene ID
696128
Gene Name
hyaluronidase 4
Gene Aliases
-
Chromosome Location
Chr.3: 150285151 - 150307095 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015134816.2 XP_014990302.1 - 1126 85

View Full Product Details