Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53B
Entrez Gene ID
694493
Gene Name
family with sequence similarity 53 member B
Gene Aliases
-
Chromosome Location
Chr.9: 125068413 - 125192881 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193710.3 NP_001180639.3 - 424 95
XM_015148393.2 XP_015003879.2 - 658 95
XM_028853249.1 XP_028709082.1 - 2416 95
XM_028853250.1 XP_028709083.1 - 442 95
GenBank mRNA JU337271.1 - - 325 95
JU477976.1 - - 538 95

View Full Product Details