Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DLK2
- Entrez Gene ID
-
700106
- Gene Name
- delta like non-canonical Notch ligand 2
- Gene Aliases
-
EGK_14936
- Chromosome Location
-
Chr.4: 125873210 - 125879819 on Build Mmul_10
- Gene Symbol
- ABCC10
- Entrez Gene ID
-
700156
- Gene Name
- ATP binding cassette subfamily C member 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 125879744 - 125904977 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV726246.1 |
- |
- |
3906 |
54 |
View Full Product Details