Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNPR
Entrez Gene ID
699098
Gene Name
synaptoporin
Gene Aliases
-
Chromosome Location
Chr.2: 119438137 - 119769061 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266609.1 NP_001253538.1 - 373 96
GenBank mRNA JU332955.1 - - 373 96
JV533599.1 - - 201 96

View Full Product Details