Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNIP2
Entrez Gene ID
712434
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 102314803 - 102343098 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194520.3 NP_001181449.1 - 834 72
XM_001111488.4 XP_001111488.1 - 2327 72
XM_001111647.3 XP_001111647.1 - 2236 72
XM_001111683.4 XP_001111683.2 - 2827 72
XM_015147988.2 XP_015003474.2 - 2837 72
XM_015147991.2 XP_015003477.1 - 1540 72
XM_015147993.2 XP_015003479.1 - 402 72
XM_028853486.1 XP_028709319.1 - 1414 72
XM_028853487.1 XP_028709320.1 - 687 72
XM_028853488.1 XP_028709321.1 - 2332 72
GenBank mRNA JU321922.1 - - 737 72
JV564982.1 - - 581 72
JV602239.1 - - 767 72
JV713381.1 - - 71
JV713383.1 - - 71

View Full Product Details