Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MZB1
Entrez Gene ID
693572
Gene Name
marginal zone B and B1 cell specific protein
Gene Aliases
EGK_15728
Chromosome Location
Chr.6: 136790620 - 136792908 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260618.1 NP_001247547.1 - 456 112
GenBank mRNA JU475860.1 - - 456 112

View Full Product Details