Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
706593
Gene Name
solute carrier family 16 member 1
Gene Aliases
EGK_01106
Chromosome Location
Chr.1: 111271542 - 111314684 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257704.1 NP_001244633.1 - 971 103
XM_015147143.2 XP_015002629.2 - 896 103
XM_015147153.2 XP_015002639.2 - 1552 103
GenBank mRNA DQ147927.1 - - 240 103
JU323021.1 - - 971 103
JU473026.1 - - 1020 103
JV721619.1 - - 979 103

View Full Product Details