Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TACR1
Entrez Gene ID
653008
Gene Name
tachykinin receptor 1
Gene Aliases
NK-1
Chromosome Location
Chr.13: 33609958 - 33757383 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037861.1 NP_001032950.1 - 528 116
GenBank mRNA AB180074.1 - - 528 116
JV703999.1 - - 965 116

View Full Product Details