Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL8
Entrez Gene ID
574179
Gene Name
C-C motif chemokine ligand 8
Gene Aliases
-
Chromosome Location
Chr.16: 30691225 - 30692607 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032851.1 NP_001028023.1 - 75 71
GenBank mRNA AF449269.1 - - 75 71

View Full Product Details