Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
BPIFA4
Entrez Gene ID
688385
Gene Name
BPI fold containing family A, member 4
Gene Aliases
BASE
Chromosome Location
Chr.10: 68407031 - 68416697 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040415.2 NP_001035505.1 - 248 110
GenBank mRNA AY913830.1 - - 250 110

View Full Product Details