Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TFB1M
- Entrez Gene ID
-
701830
- Gene Name
- transcription factor B1, mitochondrial
- Gene Aliases
-
EGK_15233
- Chromosome Location
-
Chr.4: 61552357 - 61616302 on Build Mmul_10
- Gene Symbol
- TIAM2
- Entrez Gene ID
-
701688
- Gene Name
- TIAM Rac1 associated GEF 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 61288695 - 61561905 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV704029.1 |
- |
- |
3244 |
83 |
View Full Product Details