Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP2
Entrez Gene ID
705475
Gene Name
fatty acid binding protein 2
Gene Aliases
EGK_16050
Chromosome Location
Chr.5: 117358897 - 117363775 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139143.2 XP_014994629.2 - 346 70

View Full Product Details