Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS2
Entrez Gene ID
693426
Gene Name
GINS complex subunit 2
Gene Aliases
-
Chromosome Location
Chr.20: 72708701 - 72719150 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001082775.4 XP_001082775.1 - 206 139
GenBank mRNA JU475964.1 - - 168 139

View Full Product Details