Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSR3
Entrez Gene ID
677691
Gene Name
TSR3 ribosome maturation factor
Gene Aliases
C16orf42, C20H16orf42
Chromosome Location
Chr.20: 1357073 - 1359748 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193827.3 NP_001180756.2 - 770 93
XM_028840350.1 XP_028696183.1 - 871 93
GenBank mRNA JV605230.1 - - 823 93

View Full Product Details