Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCCRP1
Entrez Gene ID
695761
Gene Name
NCCRP1, F-box associated domain containing
Gene Aliases
-
Chromosome Location
Chr.19: 38443131 - 38447647 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086619.4 XP_001086619.2 - 821 102

View Full Product Details