Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IDNK
Entrez Gene ID
708613
Gene Name
IDNK gluconokinase
Gene Aliases
EGK_07818
Chromosome Location
Chr.15: 98933301 - 98955027 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001104510.4 XP_001104510.1 - 344 81
XM_015118004.2 XP_014973490.2 - 407 81
XM_028835285.1 XP_028691118.1 - 361 81
XM_028835289.1 XP_028691122.1 - 270 81
XM_028835290.1 XP_028691123.1 - 248 81
XM_028835291.1 XP_028691124.1 - 233 81
XM_028835292.1 XP_028691125.1 - 188 81
XM_028835293.1 XP_028691126.1 - 203 81
GenBank mRNA JU343656.1 - - 190 81
JV582035.1 - - 241 81

View Full Product Details