Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
GPRIN3
Entrez Gene ID
703567
Gene Name
GPRIN family member 3
Gene Aliases
-
Chromosome Location
Chr.5: 87612617 - 87682923 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001101149.4 XP_001101149.2 - 361 134
GenBank mRNA JV704146.1 - - 324 134

View Full Product Details