Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3B
Entrez Gene ID
720683
Gene Name
outer dense fiber of sperm tails 3B
Gene Aliases
-
Chromosome Location
Chr.10: 959227 - 961407 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194710.1 NP_001181639.1 - 815 156

View Full Product Details