Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF11
Entrez Gene ID
713784
Gene Name
immunoglobulin superfamily member 11
Gene Aliases
EGK_11315
Chromosome Location
Chr.2: 157147848 - 157361970 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257906.1 NP_001244835.1 - 460 62
XM_028843355.1 XP_028699188.1 - 612 62
XM_028843356.1 XP_028699189.1 - 362 62
GenBank mRNA JU322970.1 - - 460 62
JV044976.1 - - 461 62

View Full Product Details