Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTCA
Entrez Gene ID
711623
Gene Name
RNA 3'-terminal phosphate cyclase
Gene Aliases
RTCD1
Chromosome Location
Chr.1: 123859678 - 123884587 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107617.4 XP_001107617.1 - 266 148
XM_015145898.2 XP_015001384.1 - 252 148
XM_015145905.2 XP_015001391.1 - 208 148
XM_028831210.1 XP_028687043.1 - 266 148
GenBank mRNA JV048550.1 - - 141 148
JV489142.1 - - 210 148

View Full Product Details