Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHOX2B
Entrez Gene ID
701940
Gene Name
paired like homeobox 2B
Gene Aliases
-
Chromosome Location
Chr.5: 40913901 - 40918780 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001098028.3 XP_001098028.1 - 792 124

View Full Product Details