Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A4
Entrez Gene ID
695731
Gene Name
solute carrier family 7 member 4
Gene Aliases
EGK_02814
Chromosome Location
Chr.10: 31831773 - 31835986 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266035.2 NP_001252964.1 - 1670 120
XM_028827174.1 XP_028683007.1 - 1702 120
GenBank mRNA JU335999.1 - - 1685 120
JV047983.1 - - 1671 120
JV555356.1 - - 1681 120

View Full Product Details