Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACBD6
Entrez Gene ID
716418
Gene Name
acyl-CoA binding domain containing 6
Gene Aliases
EGK_01888
Chromosome Location
Chr.1: 37476173 - 37703463 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261348.2 NP_001248277.1 - 748 109
XM_028845594.1 XP_028701427.1 - 1154 109
GenBank mRNA JU321541.1 - - 815 109
JU471387.1 - - 810 109
JV048444.1 - - 748 109
JV567350.1 - - 789 109

View Full Product Details