Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT3A
Entrez Gene ID
696200
Gene Name
Wnt family member 3A
Gene Aliases
-
Chromosome Location
Chr.1: 85583962 - 85638558 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085785.4 XP_001085785.1 - 613 75

View Full Product Details