Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFM3
Entrez Gene ID
712685
Gene Name
olfactomedin 3
Gene Aliases
EGK_01021
Chromosome Location
Chr.1: 122162088 - 122364946 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266811.1 NP_001253740.1 - 223 76
GenBank mRNA JU319935.1 - - 223 76
JV566130.1 - - 267 76
JV601984.1 - - 279 76

View Full Product Details