Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB11
Entrez Gene ID
701449
Gene Name
serpin family B member 11
Gene Aliases
EGK_09748, SERPINB8P1, SERPINBP1
Chromosome Location
Chr.18: 16403183 - 16423782 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193923.3 NP_001180852.1 - 228 110

View Full Product Details