Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIM2
Entrez Gene ID
719260
Gene Name
absent in melanoma 2
Gene Aliases
-
Chromosome Location
Chr.1: 91777410 - 91902069 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001117183.4 XP_001117183.3 - 675 127
XM_015112746.2 XP_014968232.2 - 695 127
XM_028827216.1 XP_028683049.1 - 744 127
XM_028827222.1 XP_028683055.1 - 393 127
XM_028827225.1 XP_028683058.1 - 437 127
GenBank mRNA JU325040.1 - - 353 127
JU471931.1 - - 604 127

View Full Product Details