Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLANA
Entrez Gene ID
715882
Gene Name
melan-A
Gene Aliases
EGK_07720
Chromosome Location
Chr.15: 77859791 - 77878200 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117719.2 XP_014973205.1 - 446 117

View Full Product Details