Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NNMT
Entrez Gene ID
695185
Gene Name
nicotinamide N-methyltransferase
Gene Aliases
EGK_06915
Chromosome Location
Chr.14: 107444179 - 107461226 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086727.4 XP_001086727.1 - 1268 93

View Full Product Details