Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TIMM23
- Entrez Gene ID
-
709594
- Gene Name
- translocase of inner mitochondrial membrane 23 homolog (yeast)
- Gene Aliases
-
EGK_19619
- Chromosome Location
-
Chr.9: 47388111 - 47419947 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV491241.1 |
- |
- |
444 |
139 |
View Full Product Details