The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001095865.4 | XP_001095865.1 | - | 512 | 117 |
XM_002800178.3 | XP_002800224.1 | - | 619 | 117 | |
XM_015116797.2 | XP_014972283.1 | - | 505 | 117 | |
XM_028834805.1 | XP_028690638.1 | - | 459 | 117 | |
GenBank mRNA | JU336290.1 | - | - | 120 | 117 |
JU493006.1 | - | - | 523 | 117 |