Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB44
Entrez Gene ID
715415
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
EGK_07099
Chromosome Location
Chr.14: 122888493 - 122962884 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113645.4 XP_001113645.3 - 1136 158
XM_015116125.2 XP_014971611.1 - 1618 158
XM_015116134.2 XP_014971620.1 - 1412 158
XM_028834165.1 XP_028689998.1 - 1613 158
GenBank mRNA JU321749.1 - - 1092 158
JU471366.1 - - 1119 158

View Full Product Details