Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSRC1
Entrez Gene ID
704232
Gene Name
arginine and serine rich coiled-coil 1
Gene Aliases
EGK_11960
Chromosome Location
Chr.2: 60394866 - 60806953 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266753.2 NP_001253682.1 - 536 64
XM_015131741.2 XP_014987227.1 - 704 64
XM_015131742.2 XP_014987228.1 - 673 64
XM_028843222.1 XP_028699055.1 - 740 64
GenBank mRNA JU334862.1 - - 644 64
JU475883.1 - - 645 64
JV043265.1 - - 538 64

View Full Product Details