Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F2
Entrez Gene ID
707332
Gene Name
solute carrier family 35 member F2
Gene Aliases
EGK_06858
Chromosome Location
Chr.14: 101039410 - 101104869 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260513.1 NP_001247442.1 - 872 137
XM_015115593.2 XP_014971079.1 - 856 137
XM_028832651.1 XP_028688484.1 - 790 137
GenBank mRNA JV047447.1 - - 872 137
JV636236.1 - - 794 137

View Full Product Details