Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA9
Entrez Gene ID
700612
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Gene Aliases
-
Chromosome Location
Chr.5: 39491204 - 39513567 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001094288.4 XP_001094288.1 - 3241 94
GenBank mRNA JU489371.1 - - 354 94

View Full Product Details