Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARPBP
Entrez Gene ID
696907
Gene Name
PARP1 binding protein
Gene Aliases
-
Chromosome Location
Chr.11: 101917840 - 101984180 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015152609.2 XP_015008095.2 - 1369 123
XM_015152610.2 XP_015008096.1 - 1198 123

View Full Product Details