Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DZANK1
Entrez Gene ID
700143
Gene Name
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases
C10H20orf12
Chromosome Location
Chr.10: 48008924 - 48128756 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015149557.2 XP_015005043.2 - 1574 97
XM_015149559.2 XP_015005045.2 - 1644 97
XM_028828148.1 XP_028683981.1 - 1709 97
XM_028828149.1 XP_028683982.1 - 1600 97
XM_028828150.1 XP_028683983.1 - 2109 97
XM_028828151.1 XP_028683984.1 - 2091 97
XM_028828152.1 XP_028683985.1 - 2166 97
XM_028828153.1 XP_028683986.1 - 2166 97
XM_028828154.1 XP_028683987.1 - 2043 97
XM_028828156.1 XP_028683989.1 - 2166 97

View Full Product Details