Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
713816
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
-
Chromosome Location
Chr.15: 4836749 - 4849990 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015116444.2 XP_014971930.2 - 965 69
GenBank mRNA JV690484.1 - - 878 69

View Full Product Details