Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB17
Entrez Gene ID
703609
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.12: 124964481 - 124981665 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001091918.4 XP_001091918.3 - 764 104
XM_015111418.2 XP_014966904.2 - 697 104
GenBank mRNA JV043961.1 - - 703 104
JV635425.1 - - 703 104

View Full Product Details