Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KXD1
Entrez Gene ID
720235
Gene Name
KxDL motif containing 1
Gene Aliases
C19orf50
Chromosome Location
Chr.19: 18116068 - 18126640 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194825.1 NP_001181754.1 - 381 144
XM_015123314.2 XP_014978800.1 - 480 144
XM_028838552.1 XP_028694385.1 - 504 144
GenBank mRNA JU336339.1 - - 383 144
JU477162.1 - - 443 144
JV045587.1 - - 366 144
JV529003.1 - - 405 144

View Full Product Details