Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL22
Entrez Gene ID
708739
Gene Name
methyltransferase like 22
Gene Aliases
C16orf68
Chromosome Location
Chr.20: 8673539 - 8699653 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194202.2 NP_001181131.2 - 825 108
XM_015125570.2 XP_014981056.2 - 1135 108
XM_015125571.2 XP_014981057.2 - 1148 108
XM_015125573.2 XP_014981059.2 - 1135 108
XM_028840519.1 XP_028696352.1 - 1135 108
GenBank mRNA JU323753.1 - - 1061 108
JU472906.1 - - 949 108

View Full Product Details