Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM237
Entrez Gene ID
702262
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4
Chromosome Location
Chr.12: 88928344 - 88951753 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257889.1 NP_001244818.1 - 356 150
XM_015110633.2 XP_014966119.2 - 325 150
XM_028830481.1 XP_028686314.1 - 306 150
XM_028830482.1 XP_028686315.1 - 253 150
XM_028830483.1 XP_028686316.1 - 383 150
XM_028830484.1 XP_028686317.1 - 325 150
GenBank mRNA JU335979.1 - - 356 150
JU335980.1 - - 298 150

View Full Product Details