Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM237
Entrez Gene ID
702262
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4
Chromosome Location
Chr.12: 88928344 - 88951753 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257889.1 NP_001244818.1 - 758 139
XM_015110633.2 XP_014966119.2 - 727 139
XM_028830481.1 XP_028686314.1 - 708 139
XM_028830482.1 XP_028686315.1 - 655 139
GenBank mRNA JU335979.1 - - 758 139
JU335980.1 - - 700 139

View Full Product Details