Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4E3
Entrez Gene ID
696303
Gene Name
eukaryotic translation initiation factor 4E family member 3
Gene Aliases
-
Chromosome Location
Chr.2: 127761699 - 127838709 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266831.1 NP_001253760.1 - 478 101
XM_015130398.2 XP_014985884.1 - 666 101
XM_015130399.2 XP_014985885.1 - 611 101
XM_028843118.1 XP_028698951.1 - 507 101
XM_028843119.1 XP_028698952.1 - 714 101
XM_028843120.1 XP_028698953.1 - 568 101
XM_028843121.1 XP_028698954.1 - 319 101
GenBank mRNA JU320497.1 - - 478 101
JU326026.1 - - 368 101
JV637542.1 - - 1421 101
JV703382.1 - - 478 101

View Full Product Details