Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTE3
Entrez Gene ID
114677428
Gene Name
t-complex-associated-testis-expressed 3
Gene Aliases
EGK_21332
Chromosome Location
Chr.4: 866979 - 879607 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028846951.1 XP_028702784.1 - 623 142

View Full Product Details