The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_001112428.3 | XP_001112428.1 | - | 738 | 135 |
XM_028827763.1 | XP_028683596.1 | - | 1005 | 135 | |
GenBank mRNA | JV711261.1 | - | - | 336 | 135 |